Learn More
TFA Dostmann™ Lumatic Starlight™ LED Pocket Torch
Product Code. 11802522
Change view
Click to view available options
Light Source:
LED
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Light Source Unit Size
11802522 LED Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 11802522 Supplier TFA Dostmann™ Supplier No. 43.2024
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Whether there's a power outage or for other activities in the dark, a reliable and robust torch can be a vital tool to any household. The Lumatic Starlight™ has a solid, splashproof aluminum housing and is ideal for using outdoors and at work.
  • With 51 LEDs, ultra bright
  • Up to 50 illumination range
  • Robust housing made of aluminum
  • Extreme energy saving
  • Splashproof
  • Provided with wrist strap and batteries
Specifications
Light Source LED
Weight (Metric) 200 g
Battery Type 4 x AAA 1.5 V
Height (Metric) 147 mm
Length (Metric) 57 mm
Material Aluminum
Width (Metric) 57 mm