missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide, Acid Technical A, EcoChrom™, MP Biomedicals™

Product Code. 15566226 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 kg
Unit Size:
50kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15566226 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15566226 Supplier MP Biomedicals Supplier No. 0207895
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Alumina is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1344-28-1
Color Pink to White
Physical Form Free-flowing Powder
Quantity 50 kg
Molecular Formula Al2O3
MDL Number MFCD00003424
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
IUPAC Name dialuminum;oxygen(2-)
Molecular Weight (g/mol) 101.96
PubChem CID 9989226
Formula Weight 101.96
Density 3.94g/cm3 at 20°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.