missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KinaseMax™ 5' End-Labeling Kit

Product Code. 10065024
Change view
Click to view available options
Quantity:
30 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10065024 30 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10065024 Supplier Invitrogen™ Supplier No. AM1520

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Includes

T4 Polynucleotide Kinase, Calf Intestine Alkaline Phosphatase, 10X Kinase Buffer, Dephosphorylation Buffer, Phosphatase Removal Reagent, RNA Transcript Control, pUC19/Sau3 A Markers, ATP Solution, Gel Loading Buffer II, Nuclease-free water

Produces greater yields than with standard kinase buffers

The Invitrogen KinaseMax™ 5' End-Labeling Kit allows the efficient end-labeling of DNA or RNA to high-specific activity with T4 polynucleotide kinase and [gamma-32P] ATP, or quantitative phosphorylation of 5' ends using unlabeled ATP.

The kit includes a kinase reaction buffer that exceeds the performance of the standard forward reaction buffers recommended in “Molecular Cloning: A Laboratory Manual” (Sambrook et al., 1989) and “Current Protocols in Molecular Biology” (John Wiley & Sons, Inc. Ausubel, FM et al. editors, 1994). Crude 7000 Ci/mmol [gamma-32P]ATP is compatible with this kit. End-labeled probes are more stable than internally labeled probes and can routinely be used in assays for two to four weeks.

Rapid, Phenol-Free Phosphatase Removal Step
Inactivating the Calf Intestinal Phosphatase (CIP) after the dephosphorylation reaction is a time-consuming and cumbersome procedure that usually requires phenol extraction and ethanol precipitation typically resulting in sample loss. The KinaseMax Kit now includes a novel Phosphatase Removal Reagent for quick and complete removal of CIP. After the dephosphorylation step, the reagent is added to the reaction, incubated at room temperature for 2 minutes. The tube is then spun for a minute in the microfuge and the supernatant transferred to a new tube for the kinasing reaction.

Accessory Products:
The NucAway™ Spin Columns (Cat. No. AM10070) are available for rapid purification of end-labeled probes.

  • Includes sufficient reagents for 30 reactions
  • Label oligonucleotide probes for nuclease protection assays and blot hybridizations
  • Label primers for northerns and quantitative PCR
  • Obtain up to 3-fold greater yields than with standard kinase buffers
  • Reagents included for forward and dephosphorylation reactions

DNA and RNA Purification and Analysis, DNA Labeling, Northern Blotting, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis and Blotting, Nucleic Acid Labeling and Oligo Synthesis, Oligo Labeling, RNA Labeling, Southern Blotting

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Labeling Kit
Detection Method Radioactive
Assay Nuclease Protection Assay
Content And Storage T4 Polynucleotide Kinase, Calf Intestine Alkaline Phosphatase, 10X Kinase Buffer, Dephosphorylation Buffer, Phosphatase Removal Reagent, RNA Transcript Control, pUC19⁄Sau3 A Markers, ATP Solution, Gel Loading Buffer II should be stored at –20°C. Nuclease-free water may be stored at any temperature.
Final Product Type Probes (Labeled DNA), Probes (Labeled RNA)
Label or Dye 32P (Phosphorous-32)
Includes Label or Dye No
Labeling Target cDNA, DNA (General), PCR Products, Oligos, RNA (General)
Labeling Method Direct Labeling
Shipping Condition Dry Ice
Format Kit
Product Line Ambion, KinaseMax
Quantity 30 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.