missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ Start Western Blotting Detection Reagent Kit

Product Code. 15307665 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Kit (20 mL)
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15307665 1 Kit (20 mL) 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15307665 Supplier Cytiva Supplier No. RPN3245

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemiluminescent detection of medium- and high-expression proteins, with a long lasting signal that provides an extended time window for detection.

  • Recommended for confirmatory analysis and detection of high to medium levels of endogenous proteins
  • A long signal duration enables multiple exposures of the membrane
  • Assay design flexibility and robust detection of large experimental sets
  • Produces low background, regardless of the method of detection
  • A shelf life of 18 months from manufacturing is convenient for those who run Western blots infrequently
TRUSTED_SUSTAINABILITY

Specifications

Product Type Western Blotting Detection Reagent
Kit Contents (10 mL) Luminol Solution, and (10 mL) Peroxide Solution
Membrane PVDF (Amersham Hybond P), or Nitrocellulose (Amersham Protran)
Detection Method X-ray Film or CCD Imager
Membrane Compatibility Nitrocellulose, PVDF
Quantity 1 Kit (20 mL)
For Use With (Application) Routine and Confirmatory Western Blotting
Storage Requirements 18°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.