missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMIGO3 Antibody (CL11023) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07991R
This item is not returnable.
View return policy
Description
AMIGO3 Monoclonal antibody specifically detects AMIGO3 in Human samples. It is validated for Western Blot
Specifications
| AMIGO3 | |
| Monoclonal | |
| DyLight 550 | |
| adhesion molecule with Ig-like domain 3, ALI3, Alivin-3, AMIGO-3, amphoterin-induced gene and ORF 3, amphoterin-induced protein 3, KIAA1851, MGC120552 | |
| This antibody was developed using a synthetic peptide derived from Q86WK7, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| CL11023 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 386724 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction