missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Amiloride hydrochloride

Product Code. 18732494
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18732494 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18732494 Supplier R&D Systems™ Supplier No. 0890/100
  • ONLINE EXCLUSIVE PRICE
    102.00€
    95.55€ / 100mg
    Save 6.45€
    6% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Na+ channel blocker; also I2 imidazoline ligand

Na+ channel blocker. Defines the I2A-amiloride sensitive and I2B-amiloride insensitive imidazoline binding Blocks TRPP3, acid sensing- (ASIC) and mechanogated membrane-ion channels, as well as the Na+/H+ exchanger. Also inhibits urokinase-type plasminogen activator (uPA); has no effect on tissue-type plasminogen activator.
TRUSTED_SUSTAINABILITY

Specifications

CAS 2016-88-8
Quantity 100 mg
Product Line Tocris
Molecular Formula C6H9Cl2N7O
Formula Weight 266.09
Purity >98%
Molecular Weight (g/mol) 266.09
Chemical Name or Material Amiloride hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.