missing translation for 'onlineSavingsMsg'
Learn More

Amiloride Hydrochloride, TRC

Product Code. 18041910
Change view
Click to view available options
Quantity:
50 mg
100 mg
5 g
Unit Size:
100mg
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18041910 5 g 5g
18062010 50 mg 50mg
18030910 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18041910 Supplier Toronto Research Chemicals Supplier No. TRCA5787005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2016-88-8
Molecular Formula C6 H8 Cl N7 O . Cl H
Molecular Weight (g/mol) 266.09
Synonym 2-Pyrazinecarboxamide, 3,5-diamino-N-(aminoiminomethyl)-6-chloro-, hydrochloride (1:1), Pyrazinecarboxamide, 3,5-diamino-N-(aminoiminomethyl)-6-chloro-, monohydrochloride (9CI), Pyrazinecarboxamide, N-amidino-3,5-diamino-6-chloro-, monohydrochloride (8CI), 1-(3,5-Diamino-6-chloropyrazinecarboxyl)guanidine hydrochloride, Amilorid hydrochloride, Amiloride hydrochloride, Amipramidine, Amipramizide, Amiprazide, Colectril, Guanamprazine hydrochloride, MK-870 hydrochloride, Midamor, N-Amidino-3,5-diamino-6-chloropyrazinamide hydrochloride, N-Amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride, Nilurid
IUPAC Name 3,5-diamino-N-carbamimidoyl-6-chloropyrazine-2-carboxamide;hydrochloride
SMILES Cl.NC(=N)NC(=O)c1nc(Cl)c(N)nc1N

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H8ClN7O.ClH/c7-2-4(9)13-3(8)1(12-2)5(15)14-6(10)11;/h(H4,8,9,13)(H4,10,11,14,15);1H
Formula Weight 265.0246
Chemical Name or Material Amiloride hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.