missing translation for 'onlineSavingsMsg'
Learn More

Aminophylline, anhydrous, 98%, Thermo Scientific™

A non-selective phosphodiesterase inhibitor

Brand:  Thermo Scientific Alfa Aesar J60705.14

Quantity 25g

 View more versions of this product

Product Code. 15463429

  • 46.60€ / 25g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water. Insoluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

317-34-0
420.434
FQPFAHBPWDRTLU-UHFFFAOYSA-N
9433
CN1C2=C(C(=O)N(C1=O)C)NC=N2.CN1C2=C(C(=O)N(C1=O)C)NC=N2.C(CN)N
C16H24N10O4
MFCD00013221
aminophylline, aminophyllin, theophyllamine, cardophyllin, phyllocontin, somophyllin, truphylline, theophylline ethylenediamine, cardiofilina, ethophylline
1,3-dimethyl-7H-purine-2,6-dione;ethane-1,2-diamine

Specifications

317-34-0
MFCD00013221
UN2811
aminophylline, aminophyllin, theophyllamine, cardophyllin, phyllocontin, somophyllin, truphylline, theophylline ethylenediamine, cardiofilina, ethophylline
FQPFAHBPWDRTLU-UHFFFAOYSA-N
1,3-dimethyl-7H-purine-2,6-dione;ethane-1,2-diamine
9433
98%
C16H24N10O4
25g
14,465
Soluble in water. Insoluble in alcohol and ether.
CN1C2=C(C(=O)N(C1=O)C)NC=N2.CN1C2=C(C(=O)N(C1=O)C)NC=N2.C(CN)N
420.434
210.21
Aminophylline, anhydrous
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.