missing translation for 'onlineSavingsMsg'
Learn More

Aminophylline, anhydrous, 98%, Thermo Scientific™

A non-selective phosphodiesterase inhibitor

46.60€ - 97.00€

Chemical Identifiers

CAS 317-34-0
Molecular Formula C16H24N10O4
Molecular Weight (g/mol) 420.434
MDL Number MFCD00013221
InChI Key FQPFAHBPWDRTLU-UHFFFAOYSA-N
Synonym aminophylline, aminophyllin, theophyllamine, cardophyllin, phyllocontin, somophyllin, truphylline, theophylline ethylenediamine, cardiofilina, ethophylline
PubChem CID 9433
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione;ethane-1,2-diamine
SMILES CN1C2=C(C(=O)N(C1=O)C)NC=N2.CN1C2=C(C(=O)N(C1=O)C)NC=N2.C(CN)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15463429
SDS View Documents
Thermo Scientific Alfa Aesar
J60705.14
25g
46.60€
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15473429
SDS View Documents
Thermo Scientific Alfa Aesar
J60705.22
100g
97.00€
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water. Insoluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents.
Specifications

Chemical Identifiers

317-34-0
420.434
FQPFAHBPWDRTLU-UHFFFAOYSA-N
9433
CN1C2=C(C(=O)N(C1=O)C)NC=N2.CN1C2=C(C(=O)N(C1=O)C)NC=N2.C(CN)N
C16H24N10O4
MFCD00013221
aminophylline, aminophyllin, theophyllamine, cardophyllin, phyllocontin, somophyllin, truphylline, theophylline ethylenediamine, cardiofilina, ethophylline
1,3-dimethyl-7H-purine-2,6-dione;ethane-1,2-diamine

Specifications

317-34-0
MFCD00013221
14,465
Soluble in water. Insoluble in alcohol and ether.
CN1C2=C(C(=O)N(C1=O)C)NC=N2.CN1C2=C(C(=O)N(C1=O)C)NC=N2.C(CN)N
420.434
210.21
Aminophylline, anhydrous
C16H24N10O4
UN2811
aminophylline, aminophyllin, theophyllamine, cardophyllin, phyllocontin, somophyllin, truphylline, theophylline ethylenediamine, cardiofilina, ethophylline
FQPFAHBPWDRTLU-UHFFFAOYSA-N
1,3-dimethyl-7H-purine-2,6-dione;ethane-1,2-diamine
9433
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 206-264-5

missing translation for 'rtecsNumber' : XH5600000

missing translation for 'tsca' : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.