missing translation for 'onlineSavingsMsg'
Learn More

Aminopyrine (N,N-Dimethyl-13C2), TRC

Product Code. 30135550
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135550 250 mg 250mg
30133708 100 mg 100mg
30011355 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135550 Supplier Toronto Research Chemicals Supplier No. TRCA639995250MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60433-90-1
Molecular Formula C1113C2H17N3O
Molecular Weight (g/mol) 233.28
Synonym 4-[Di(methyl-13C)amino]-1,2-dihydro-1,5-dimethyl-2-phenyl-3H-Pyrazol-3-one;
IUPAC Name 4-[di((113C)methyl)amino]-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=C(N([13CH3])[13CH3])C(N(C2=CC=CC=C2)N1C)=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H17N3O/c1-10-12(14(2)3)13(17)16(15(10)4)11-8-6-5-7-9-11/h5-9H,1-4H3/i2+1,3+1
Formula Weight 233.1439
Percent Purity >95%
Chemical Name or Material Aminopyrine (N,N-Dimethyl-13C2)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.