missing translation for 'onlineSavingsMsg'
Learn More

Amisulpride, TRC

Product Code. 17924165
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17924165 50 mg 50mg
17993815 10 mg 10mg
17971195 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17924165 Supplier Toronto Research Chemicals Supplier No. TRCA63325050MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71675-85-9
Molecular Formula C17 H27 N3 O4 S
Molecular Weight (g/mol) 369.48
Synonym Amisulpride, 4-Amino-N-[[(2RS)-1-ethylpyrrolidin-2-yl]methyl]-5-(ethylsulfonyl)-2-methoxybenzamide
IUPAC Name 4-amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-5-ethylsulfonyl-2-methoxybenzamide
SMILES CCN1CCCC1CNC(=O)c2cc(c(N)cc2OC)S(=O)(=O)CC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H27N3O4S/c1-4-20-8-6-7-12(20)11-19-17(21)13-9-16(25(22,23)5-2)14(18)10-15(13)24-3/h9-10,12H,4-8,11,18H2,1-3H3,(H,19,21)
Formula Weight 369.1722
Percent Purity >95%
Chemical Name or Material Amisulpride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.