missing translation for 'onlineSavingsMsg'
Learn More

Melford Ammonium Acetate

Product Code. 30018016
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018016 5 kg 5kg
30018015 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018016 Supplier Melford Supplier No. A206005000.0
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Widely used reagent in molecular biology and chromatography.

Widely used reagent in molecular biology and chromatography.

Specifications

Chemical Name or Material Ammonium Acetate
CAS 631-61-8
Quantity 5 kg
Molecular Formula C2H7NO2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.