Learn More
Ammonium Carbonate solution EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA
Product Code. 12917163 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12917163 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12917163 Supplier Fisher Chemical Supplier No. RP 013 1000
  • ONLINE EXCLUSIVE PRICE
    115.00€
    108.82€ / 1000mL
    Save 6.18€
    5% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Packaging Bottle
Analyte or Component Names Ammonium Carbonate Solution
Purity Grade Notes EU Ph Ref: 1005201
Element Ammonium Carbonate Solution
Quantity 1000 mL
Grade Eu. Ph.
Product Identifier
  • Ammonium Carbonate Solution. European Pharmacopoeia Ref: 1005201 (RP 013)