missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) citrate, 20.5-22.5%, Specified reagent for general laboratory work, Fisher Chemical

Product Code. 11423513 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423513 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11423513 Supplier Fisher Chemical Supplier No. A/4640/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1185-57-5
Color Brown
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula C6H8FeNO7
MDL Number 13099
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-K
SMILES N.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
IUPAC Name iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate amine
Molecular Weight (g/mol) 261.98
Formula Weight 265g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.