missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Ammonium Sulfate
Description
Useful for precipitation or fractionation of proteins or for purification of antibodies.
Useful for precipitation or fractionation of proteins or for purification of antibodies. Used in crystallographic analysis of nucleic acids and proteins.
Specifications
Specifications
| Chemical Name or Material | Ammonium Sulfate |
| CAS | 7783-20-2 |
| Quantity | 1 kg |
| Molecular Formula | H8N2O4S |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?