missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetraphenylborate, 99%, Thermo Scientific™

Product Code. 15422028
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422028 25 g 25g
15412028 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422028 Supplier Thermo Scientific Alfa Aesar Supplier No. H55700.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14637-34-4
Molecular Formula C24H24BN
Molecular Weight (g/mol) 337.273
MDL Number MFCD00134523
InChI Key ZWFYDLYRTYMBIL-UHFFFAOYSA-O
Synonym ammonium tetraphenylborate, tetraphenylboron ammonium, ammonium tetraphenylborate 1-
PubChem CID 14496924
IUPAC Name azanium;tetraphenylboranuide
SMILES [B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[NH4+]

Specifications

Melting Point 220°C (decomposition)
Linear Formula NH4B(C6H5)4
Quantity 25 g
Beilstein 3799683
Formula Weight 337.27
Percent Purity 99%
Chemical Name or Material Ammonium tetraphenylborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.