missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiosulfate, Extra Pure, SLR, Fisher Chemical

Product Code. 10409170 Shop All Fisher Chemical Products
Click to view available options
:
500g, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10409170

Brand: Fisher Chemical A/6720/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 150°C
CAS 7783-18-8
Color White
pH 6.2 to 7.0
Physical Form Solid
Quantity 500 g
Molecular Formula H8N2O3S2
MDL Number 15965
InChI Key XYXNTHIYBIDHGM-UHFFFAOYSA-N
SMILES N.N.OS(O)(=O)=S
IUPAC Name dihydroxy-1λ⁶-disulfen-1-one diamine
Molecular Weight (g/mol) 148.20
Formula Weight 148.21g/mol
Packaging Plastic powder jar
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.