Learn More
Ammonium tungsten oxide pentahydrate, 99.999% (metals basis), Thermo Scientific™, Puratronic™
Product Code. 11317077
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317077 25 g 25g
11327077 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11317077 Supplier Thermo Scientific Alfa Aesar Supplier No. 010899.14
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1311-93-9
Molecular Formula (NH4)10W12O41·5H2O
MDL Number MFCD00015956
Specifications
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Slightly soluble in water.
Formula Weight 3132.64 (3042.56 Anhydrous)
Odor Odorless
Appearance White crystalline
Density 2.3 g/cm3
Chemical Name or Material Ammonium tungsten oxide pentahydrate