missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin Dimer Sodium (>90%) (Mixture of Diastereomers), TRC

Product Code. 30137257
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137257 5 mg 5mg
30007443 1 mg 1mg
30011634 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137257 Supplier Toronto Research Chemicals Supplier No. TRCA6345105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34642-77-8
Molecular Formula C32H38N6O10S2 (x Na)
Molecular Weight (g/mol) 387.4
Synonym (2R)-(2R)-2-(4-Hydroxyphenyl)glycyl-(2S)-2-[(2R,4S)-4-carboxy-5,5-dimethyl-2-thiazolidinyl]glycyl-N-[(2S,5R,6R)-2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-6-yl]-2-(4-hydroxyphenyl)glycinamide Sodium Salt, [2(2R-trans),3[2S-(2α,5α,6β)]]-D-2-(4-hydroxyphenyl)glycyl-L-2-(4-carboxy-5,5-dimethyl-2-thiazolidinyl)glycyl-N-(2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-6-yl)-D-2-(4-hydroxyphenyl)glycinamide Sodium Salt, Amoxicillin USP Related Compound J Sodium Salt
IUPAC Name (2R)-(2R)-2-(4-Hydroxyphenyl)glycyl-(2S)-2-[(2R,4S)-4-carboxy-5,5-dimethyl-2-thiazolidinyl]glycyl-N-[(2S,5R,6R)-2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-6-yl]-2-(4-hydroxyphenyl)glycinamide Sodium Salt; [2(2R-trans),3[2S-(2α,5α,6β)]]-D-2-(4-hydroxyphenyl)glycyl-L-2-(4-carboxy-5,5-dimethyl-2-thiazolidinyl)glycyl-N-(2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-6-yl)-D-2-(4-hydroxyphenyl)glycinamide Sodium Salt; Amoxicillin USP Related Compound J Sodium Salt
SMILES O=C(NC(C(N[C@H](C1=CC=C(O)C=C1)C(N[C@]([C@@]2(N[C@@H](C(O[Na])=O)C(C)(C)S2)[H])([H])C(O[Na])=O)=O)=O)[C@]3([H])N[C@@H](C(O[Na])=O)C(C)(C)S3)C(N)C4=CC=C(O)C=C4

Specifications

Analyte or Component Names Amoxicillin Dimer Sodium Salt
Shipping Condition Blue Ice
InChI Formula InChI=1S/C32H38N6O10S2/c1-31(2)21(29(45)46)37-26(49-31)19(35-23(41)17(33)13-5-9-15(39)10-6-13)25(43)34-18(14-7-11-16(40)12-8-14)24(42)36-20-27(44)38-22(30(47)48)32(3,4)50-28(20)38/h5-12,17-22,26,28,37,39-40H,33H2,1-4H3,(H,34,43)(H,35,41)(H,36,42)(H,45,46)(H,47,48)/t17-,18-,19+,20-,21+,22+,26-,28-/m1/s1
Solution Type Neat
Industry Type Forensics
Quantity 5 mg
Formula Weight 387.0865
Chemical Name or Material Amoxicillin Dimer Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.