missing translation for 'onlineSavingsMsg'
Learn More

Amylase alpha Mouse anti-Human, Porcine, Rat, Clone: 2F1G8, Proteintech

Mouse Monoclonal Antibody

145.00€ - 410.00€

Specifications

Antigen Amylase alpha
Clone 2F1G8
Concentration 0.79 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16837735
View Documents
Proteintech
66133-1-IG-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16827735
View Documents
Proteintech
66133-1-IG-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody can recognize amylase alpha.

Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. It is an amylase isoenzyme produced by the salivary gland.
Specifications

Specifications

Amylase alpha
0.79 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
AMY2A, Amylase, alpha, Pancreatic alpha amylase
AMY2A, Amy2a3
IgG2b
Protein A
Antibody
2F1G8
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
Unconjugated
Mouse
Pig, Human, Rat
P04746
100521789, 279, 497039
Amylase, alpha Fusion Protein Ag8819
Primary
-20°C
AMY2A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.