Learn More
Thermo Scientific™ AnaeroBox Pot, Rectangular Lid
Product Code. 15970527
Change view
Click to view available options
Includes:
Rectangular Lid
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
15970527 Rectangular Lid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15970527 Supplier Thermo Scientific™ Supplier No. AB0025L
  • ONLINE EXCLUSIVE PRICE
    74.00€
    67.60€ / Pack of 1
    Save 6.40€
    8% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AnaeroBox pot, rectangular (2.5 L) lid
AnaeroBox pot, rectangular (2.5 L) lid
TRUSTED_SUSTAINABILITY
Specifications
Description 2.5 L Container Lid
Includes Rectangular Lid