Learn More
Kern Sohn Analytical balance ADB
Leading analytical balance with internal adjustment.
775.20€ - 928.20€
Specifications
Readability 0.1 mg
Linearity 0.4 mg
Repeatability 0.2 mg
Electrical Requirements Universal Mains Adapter
View More Specs
Products 3
Product Code Brand Capacity Model Price Quantity & Availability  
Product Code Brand Capacity Model Price Quantity & Availability  
17301543
View Documents Promotion Details
Kern Sohn
ADB 100-4
120 g ADB
785.40€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
17311543
View Documents Promotion Details
Kern Sohn
ADB 200-4
220 g ADB
928.20€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
17321543
View Documents Promotion Details
Kern Sohn
ADB 600-C3
120 g ADB Compact
775.20€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Description
Description
  • Adjusting program CAL for quick setting of the balance accuracy using an external test weight
  • ADB 600-C3: Compact, space-saving carat balance with a readout of 0.001 ct and a weighing range of 600 ct. The high level of accuracy saves hard cash wherever you are weighing valuable precious stones
  • Level indicator and levelling feet for precise levelling of the scale, fitted as standard, to give the most accurate weighing result
  • Large glass draught shield with 3 sliding doors for easy access to the items being weighed.
  • Compact size, practical for small spaces
  • Simple and convenient 6-key operation
Specifications
Specifications
0.1 mg
0.4 mg
0.2 mg
Universal Mains Adapter