Learn More
Motic™ Analyzer Slider, with Rotating Mount, BA400/BA410
Product Code. 11726606
Change view
Click to view available options
For Use With (Equipment):
BA400 and BA410 Series Upright Compound Microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11726606 BA400 and BA410 Series Upright Compound Microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11726606 Supplier Motic™ Supplier No. 1101001900791
  • ONLINE EXCLUSIVE PRICE
    123.00€
    110.13€ / Pack of 1
    Save 12.87€
    10% Off
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Improve image contrast with a simple polarization set containing a polarizer and analyzer. Motic™ Analyzer Slider, with Rotating Mount, BA400/BA410 is rotatable in 360°.
  • Improve image quality obtained with birefringent materials when compared to other techniques such as darkfield and brightfield illumination, differential interference contrast, phase contrast, Hoffman modulation contrast, and fluorescence.
  • Useful for both quantitative and qualitative studies targeted at a wide range of anisotropic specimens.
  • Polarized light microscopy is capable of providing information on absorption color and optical path boundaries between minerals of differing refractive indices, in a manner similar to brightfield illumination, but the technique can also distinguish between isotropic and anisotropic substances.
  • Contrast-enhancing technique exploits the optical properties specific to anisotropy and reveals detailed information concerning the structure and composition of materials that are invaluable for identification and diagnostic purposes.
Specifications
Type Analyzer Slider
For Use With (Equipment) BA400 and BA410 Series Upright Compound Microscopes
Includes Rotating Mount