missing translation for 'onlineSavingsMsg'
Learn More

Anastrozole, TRC

Product Code. 30040842
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040842 50 mg 50mg
18080020 10 mg 10mg
18063700 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040842 Supplier Toronto Research Chemicals Supplier No. TRCA63742550MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120511-73-1
Molecular Formula C17 H19 N5
Molecular Weight (g/mol) 293.37
Synonym Anastrozole, 2,2'-[5-(1H-1,2,4-Triazol-1-ylmethyl)benzene-1,3-diyl]bis(2-methylpropanenitrile), 1,3-Benzenediacetonitrile, α,α,α',α'-tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)- (9CI), α1,α1,α3,α3-Tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile, 2,2'-[5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-phenylene]di(2-methylpropionitrile), 2-[3-(1-Cyano-1-methylethyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]phenyl]-2-methylpropanenitrile, 2-[3-(2-Cyano-2-propyl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-2-methylpropiononitrile, 2-[3-(2-Cyanopropan-2-yl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-2-methylpropanenitrile, Anastrazole, Anastrol, Arimidex, ICI-D 1033, ZD 1033
IUPAC Name 2-[3-(2-cyanopropan-2-yl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-2-methylpropanenitrile
SMILES CC(C)(C#N)c1cc(Cn2cncn2)cc(c1)C(C)(C)C#N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H19N5/c1-16(2,9-18)14-5-13(8-22-12-20-11-21-22)6-15(7-14)17(3,4)10-19/h5-7,11-12H,8H2,1-4H3
Formula Weight 293.164
Percent Purity >95%
Chemical Name or Material Anastrozole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.