Learn More
Abeba™ Anatom 2616 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 2616/47
Additional Details : Weight : 1.26600kg
 View more versions of this product
Product Code. 15608187
  • 108.12€ / 1 pair
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S1 SRC
  • Microfiber, perforated
  • White
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Adjustable instep area
  • Double velcro fastening
  • Reflective material
  • Reflective material
  • Suitable for hygienically demanding areas
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRC
Microfiber
Slip-resistant
2616
White
47
Polyurethane
Steel