Learn More
Abeba™ Anatom 32130 Shoes
Product Code. 15280925
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15280925 36 1 pair
15630355 37 1 pair
15200935 38 1 pair
15650355 39 1 pair
15660355 40 1 pair
15230935 41 1 pair
15680355 42 1 pair
15690355 43 1 pair
15260935 44 1 pair
15610365 45 1 pair
15620365 46 1 pair
15290935 47 1 pair
15640365 48 1 pair
15200945 48 1 pair
15210945 49 1 pair
15220945 50 1 pair
15607678 51 1 pair
15617678 52 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Product Code. 15280925 Supplier Abeba™ Supplier No. 32130/36
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber
  • White
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • velcro fastening
  • Suitable for heel and foot lift
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRC
Color White
Material Microfiber
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32130
Toe Material Steel