Learn More
IKA™ Anchor Stirrer with PEEK Scrapers
Shop All IKA Products
Click to view available options
Capacity (Metric):
400 mL min.
Unit Size:
Each
Description
- Minimum usable volume when homogenizing: 400 mL
- Stainless steel
- For use with Lab reactors
Specifications
Specifications
| Capacity (Metric) | 400 mL min. |
| Color | White |
| Includes | PEEK scapers |
| Model | LR 1000.10 |
| Shape | Anchor |
| Type | Overhead Stirrer Shaft |