missing translation for 'onlineSavingsMsg'
Learn More

Anidulafungin, TRC

Product Code. 30007427
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007427 100 mg 100mg
30005093 5 mg 5mg
30012675 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007427 Supplier Toronto Research Chemicals Supplier No. TRCA673000100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 166663-25-8
Synonym Echinocandin B, 1-[(4R,5R)-4,5-dihydroxy-N2-[[4' '-(pentyloxy)[1,1':4',1' '-terphenyl]-4-yl]carbonyl]-L-ornithine]-∣1-[(4R,5R)-4,5-Dihydroxy-N2-[[4' '-(pentyloxy)[1,1':4',1' '-terphenyl]-4-yl]carbonyl]-L-ornithine]echinocandin B∣Anidulafungin∣Ecalta∣Eraxis∣LY 303366

Specifikationer

Purity >95% (HPLC)
Format Neat
Molecular Weight (g/mol) 1140.24
Product Type API
Quantity 100 mg
Shipping Condition Room Temperature
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.