missing translation for 'onlineSavingsMsg'
Learn More

Anidulafungin, TRC

Product Code. 30012675
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012675 10 mg 10mg
30007427 100 mg 100mg
30005093 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012675 Supplier Toronto Research Chemicals Supplier No. TRCA67300010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166663-25-8
Synonym Echinocandin B, 1-[(4R,5R)-4,5-dihydroxy-N2-[[4' '-(pentyloxy)[1,1':4',1' '-terphenyl]-4-yl]carbonyl]-L-ornithine]-∣1-[(4R,5R)-4,5-Dihydroxy-N2-[[4' '-(pentyloxy)[1,1':4',1' '-terphenyl]-4-yl]carbonyl]-L-ornithine]echinocandin B∣Anidulafungin∣Ecalta∣Eraxis∣LY 303366

Specifications

Purity >95% (HPLC)
Format Neat
Molecular Formula C58H73N7O17
MDL Number MFCD00917070
Molecular Weight (g/mol) 1140.24, 1140.25
InChI Key JHVAMHSQVVQIOT-UEHAGLDDSA-N
SMILES [H][C@@]12C[C@@H](O)CN1C(=O)[C@@]([H])(NC(=O)[C@H](C[C@@H](O)[C@@H](O)NC(=O)[C@]1([H])[C@@H](O)[C@@H](C)CN1C(=O)[C@@]([H])(NC(=O)[C@@]([H])(NC2=O)[C@H](O)[C@@H](O)C1=CC=C(O)C=C1)[C@@H](C)O)NC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(OCCCCC)C=C1)[C@@H](C)O
IUPAC Name N-[(3S,6S,9S,11R,15S,18S,20R,21R,24S,25S,26S)-6-[(1S,2S)-1,2-dihydroxy-2-(4-hydroxyphenyl)ethyl]-11,20,21,25-tetrahydroxy-3,15-bis[(1R)-1-hydroxyethyl]-26-methyl-2,5,8,14,17,23-hexaoxo-1,4,7,13,16,22-hexaazatricyclo[22.3.0.0⁹,¹³]heptacosan-18-yl]-4-[4'-(pentyloxy)-[1,1'-biphenyl]-4-yl]benzamide
Product Type API
Quantity 10 mg
Shipping Condition Room Temperature
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.