missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anillin Antibody (CL0301) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52907JF646
This item is not returnable.
View return policy
Description
Anillin Monoclonal antibody specifically detects Anillin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Anillin | |
| Monoclonal | |
| Janelia Fluor 646 | |
| actin-binding protein anillin, ANILLIN, anillin (Drosophila Scraps homolog), actin binding protein, anillin, actin binding protein, anillin, actin binding protein (scraps homolog, Drosophila), DKFZp779A055, Drosophila Scraps homolog, scra, Scraps | |
| This antibody was developed using a recombinant protein derived from Q9NQW6, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL0301 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 54443 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction