Learn More
Laboratoires Anios™ Aniospray Quick Spray Surface Disinfectant
Product Code. 15451884
Change view
Click to view available options
Volume (Metric):
1 L
5 L
Quantity:
12/Cs.
4/Cs.
Unit Size:
Pack of 12
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Quantity unitSize
15451884 1 L 12/Cs. Pack of 12
15461884 5 L 4/Cs. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15451884 Supplier Laboratoires Anios™ Supplier No. 2084073
  • ONLINE EXCLUSIVE PRICE
    381.20€
    242.03€ / Pack of 12
    Save 139.17€
    36% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
Laboratoires Anios™ Aniospray Quick Spray Surface Disinfectant is an ethanol-based, broad antimicrobial spectrum spray for rapid disinfection of surfaces and non-immersible, non-invasive equipment.
  • Ethanol-based composition
  • Broad activity spectrum
  • Leaves no trace when drying
  • Pleasant fragrance
  • Aldehyde free, without coloring
  • Rapid action disinfection of surfaces and previously cleaned, non-immersible medical devices resistant to alcohol (stethoscopes, pressure sensors, blood sugar testers, etc.)
  • Active against bacteria, mycobacteria, yeasts, molds, viruses
Specifications
Product Type Rapid Disinfectant
Certifications/Compliance Bacteria: EN 13727, EN 1276, EN 13697, MRB according to EN 13697Mycobacteria: EN 14348, EN 14563Yeasts/Molds: EN 13624, EN 1650, EN 13697, EN 14562 : Tricophyton mentagrophytesViruses: Adenovirus, HIV-1, PRV (surrogate of HBV), BVDV (surrogate of HCV), Vaccinia virus Herpes-virus, Coronavirus, Rotavirus, Infl uenza virus A (H N ), Feline Calicivirus, Norovirus MNV according to EN 14476
Composition Hydroalcoholic solution (ethanol 55%), quaternary ammonium propionate,fragrance.
For Use With (Application) Rapid disinfection of non-immersible medical devices resistant to alcohol.
Quantity 12/Cs.
Volume (Metric) 1 L
compliance-icons compliance-icons
Product Identifier
  • ANIOSPRAY QUICK
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ETHYL ALCOHOL, PROPAN-2-OL, DIDECYLDIMETHYLAMMONIUM CHLORIDE