missing translation for 'onlineSavingsMsg'
Learn More

Anisaldehyde Dimethyl Acetal, TRC

Product Code. 30335710
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335710 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335710 Supplier Toronto Research Chemicals Supplier No. TRCA6732055G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Anisaldehyde Dimethyl Acetal
CAS 2186-92-7
Molecular Formula C10H14O3
Quantity 5 g
Synonym 1-(Dimethoxymethyl)-4-methoxybenzene, (p-Methoxyphenyl)dimethoxymethane, 4-Anisaldehyde Dimethyl Acetal, 4-Methoxybenzaldehyde Dimethyl Acetal, Anisaldehyde dimethyl Acetal, Anisic aldehyde Dimethyl Acetal, Dimethoxy(4-methoxyphenyl)methane, p-(Dimethoxymethyl)anisole, p-Methoxybenzaldehyde Dimethyl Acetal
InChI Formula InChI=1S/C10H14O3/c1-11-9-6-4-8(5-7-9)10(12-2)13-3/h4-7,10H,1-3H3
SMILES O=C(N([H])[C@H](C(N)=O)C(C)C)C1=C2C=CC=CC2=NN1CC3CCCCC3
IUPAC Name 1-(dimethoxymethyl)-4-methoxybenzene
Molecular Weight (g/mol) 182.22
Formula Weight 182.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.