missing translation for 'onlineSavingsMsg'
Learn More

Anisindione, TRC

Product Code. 30137256
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137256 5 g 5g
30137580 10 g 10g
30008499 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137256 Supplier Toronto Research Chemicals Supplier No. TRCA6734805G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-37-3
Molecular Formula C16 H12 O3
Molecular Weight (g/mol) 252.26
Synonym 2-(4-Methoxyphenyl)-1H-Indene-1,3(2H)-dione, 2-(p-Methoxyphenyl)-1,3-indandione, 2-p-Anisyl-1,3-indandione, Andion, Miradon, SPE 2792, Unidone
IUPAC Name 2-(4-methoxyphenyl)indene-1,3-dione
SMILES COc1ccc(cc1)C2C(=O)c3ccccc3C2=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H12O3/c1-19-11-8-6-10(7-9-11)14-15(17)12-4-2-3-5-13(12)16(14)18/h2-9,14H,1H3
Formula Weight 252.0786
Chemical Name or Material Anisindione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.