missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anisole-d3 (methyl-d3), CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Anisole (Methoxy-D3) |
| CAS | 4019-63-0 |
| Molecular Formula | C7 D3 H5 O |
| Quantity | 1 g |
| Synonym | Benzene, methoxy-d3- (9 CI, ACI), Anisole-α,α,α-d3 (6 CI, 7 CI, 8 CI), Methoxy-d3benzene (ACI), (Trideuteriomethoxy)benzene, Methoxy-d3-benzene, Anisole-d3 (methyl-d3), Anisole-d3 (methoxy-d3), Anisole-α,α,α-D3 |
| InChI Formula | InChI=1 S/C7H8O/c1-8-7-5-3-2-4-6-7/h2-6 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1ccccc1 |
| IUPAC Name | trideuteriomethoxybenzene |
| Molecular Weight (g/mol) | 111.1563 |
| Formula Weight | 111.0763 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction