missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Anisomycin ReadyMade™ Solution

Product Code. 30044022
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 0.5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30044022 10 x 0.5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044022 Supplier TOKU-E™ Supplier No. A22610x0.5ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Anisomycin ReadyMade™ Solution is provided as a filter-sterilized solution of Anisomycin dissolved in PBS.

  • CAS Number: 22862-76-6
  • Molecular Formula: C14H19NO4
  • Molecular Weight: 265.30 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Streptomyces griseolus
  • Spectrum: Anisomycin is active against Gram-negative bacteria and protozoa. It has in vitro activity against dengue and Zika flaviviruses in cell culture
  • Mechanism of Action: Anisomycin inhibits protein synthesis and depresses the level of the conventional early mRNAs by inhibiting peptidyl transferase or the 80S ribosome system in eukaryotic systems

Specifications

Chemical Name or Material Anysomicin, Flagecidin
Physical Form Liquid
CAS 22862-76-6
pH 4.6
Source Streptomyces griseolus
Packaging 2 mL plastic vial
Sterile Filtered Yes, 0.22 μm
Sterility Sterile
Color Colorless
Concentration 10.0 to 10.5 mg/mL
Quantity 10 x 0.5 mL
Purity Grade Notes >98% (HPLC)
Assay Percent Range >98.0%
Assay Percent Notes Anisomycin, Dried Basis
Molecular Formula C14H19NO4
Product Line ReadyMade™ Solutions
Shipping Condition Ice Pack
Synonym Flagecidin
IUPAC Name (2R,3S,4S)-2-(4-Methoxybenzyl)-3,4-pyrrolidinediol-3-acetate
Molecular Weight (g/mol) 265.30 g/mol
Formula Weight 265.30 amu
Percent Purity >98%
Grade Antibiotic
Concentration or Composition (by Analyte or Components) Anisomycin 1%, Phosphate Buffered Saline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.