missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKLE2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26293-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined.Specifications
| ANKLE2 | |
| Polyclonal | |
| Unconjugated | |
| ANKLE2 | |
| ANKLE2, KIAA0692, LEM4, LEMD7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23141 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86XL3 | |
| ANKLE2 | |
| ANKLE2 Fusion Protein Ag23387 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction