missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD33 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25202-1-AP-150UL
This item is not returnable.
View return policy
Description
Ankyrin repeat domain 33 (ANKRD33), also named as C12orf7, belongs to the ankyrin repeat family and is highly expressed in retina. ANKRD33 exists as two isoforms. The isoform 1 molecular weight is 29 kDa and isoform 2 is 49 kDa. The function of ANKRD33 is still non-known.Specifications
| ANKRD33 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD33 | |
| ANKRD33, ankyrin repeat domain 33, C12orf7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 341405 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7Z3H0 | |
| ANKRD33 | |
| ANKRD33 Fusion Protein Ag18582 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction