missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin, bacteria-like protein Antibody (V71), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59672PCP
This item is not returnable.
View return policy
Description
Peroxiredoxin, bacteria-like protein Monoclonal antibody specifically detects Peroxiredoxin, bacteria-like protein in Parasite samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| Peroxiredoxin, bacteria-like protein | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Peptide Sequence specific to peroxiredoxin antigen from Tetracapsuloides bryosalmonae (fish parasite). | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| V71 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Parasite | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction