missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584635
This item is not returnable.
View return policy
Description
Immunogen sequence: TGYPPFPGYP PAGQESSFPP SGQYPYPSGF PPMGGGAYPQ VPSSGYPGAG GYPAP Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.
Calcium/phospholipid-binding protein which promotes membrane fusion and is involved in exocytosis. Annexin VII is a member of the annexin family of calcium-dependent phospholipid binding proteins. The Annexin VII gene contains 14 exons and spans approxima.
Specifications
| Annexin A7 | |
| Polyclonal | |
| Unconjugated | |
| ANXA7 | |
| AI265384; AI316497; Annexin A7; annexin VII; Annexin-7; Anx7; ANXA7; Interleukin-8; OK/SW-cl.95; RP11-537A6.8; SNX; SYNEXIN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 310 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20073 | |
| ANXA7 | |
| Recombinant protein corresponding to Human ANXA7. Recombinant protein control fragment (Product #RP-105306). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction