Learn More
Invitrogen™ Annexin V Monoclonal Antibody (VAA-33), FITC, eBioscience™

Mouse Monoclonal Antibody
Brand: Invitrogen™ BMS147FI
Description
Description: BMS147 specifically recognizes Annexin V (VAC- alpha ). BMS147 is suitable for ELISA application, Western blot analysis and is suitable as primary antibody for staining in FACS analysis. BMS147FI can be used for direct staining of cells. BMS147BT can be used in a two-step procedure using the biotin-(strept-)avidin system. For intracellular stainings the use of BMS147FI is recommended. Applications Tested: Flow Cytometry (Intracellular Staining). Purity: >95%. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
Specifications
| Annexin V | |
| Monoclonal | |
| 10 μL/Test | |
| TBS with 1% BSA and 0.02% sodium azide; pH 7.2-7.4 | |
| P08758 | |
| Anxa5 | |
| Affinity chromatography | |
| RUO | |
| 308 | |
| 4°C, store in dark | |
| Liquid | |
| Responsibly packaged |
| Flow Cytometry | |
| VAA-33 | |
| FITC | |
| Anxa5 | |
| Anchorin CII; annexin 5; annexin A5; Annexin V; annexin-5; Anx5; ANXA5; Calphobindin I; CBP-I; Endonexin II; ENX2; epididymis secretory protein Li 7; HEL-S-7; LC5; lipocortin V; PAP-I; placental anticoagulant protein 4; Placental anticoagulant protein I; PP4; R74653; RPRGL3; Thromboplastin inhibitor; VAC-alpha; Vascular anticoagulant-alpha | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.