missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Annexin V Recombinant Protein, FITC, eBioscience™

Product Code. 15531917
Change view
Click to view available options
Quantity:
100 Tests
20 Tests
300 Tests
Unit Size:
100 tests
20 tests
300 tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15531917 20 Tests 20 tests
15521917 100 Tests 100 tests
15541917 300 Tests 300 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15531917 Supplier Invitrogen™ Supplier No. BMS306FI20
  • ONLINE EXCLUSIVE PRICE
    154.00€
    115.65€ / 20 tests
    Save 38.35€
    24% Off
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Recombinant Protein

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08758, P14668, P48036
Quantity 20 Tests
Concentration 5 μL/Test
For Use With (Application) Neutralization, Flow Cytometry
Formulation TBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene ID (Entrez) 11747, 25673, 308
Molecular Weight (g/mol) 35.8kDa
Name Annexin V
pH Range 7.4
Purification Method Purified
Source E. coli
Storage Requirements 4°C, store in dark, DO NOT FREEZE!
Regulatory Status RUO
Structural Form E. Coli
Gene Alias Anchorin CII; annexin 5; annexin A5; Annexin V; annexin-5; Anx5; ANXA5; Calphobindin I; CBP-I; Endonexin II; ENX2; epididymis secretory protein Li 7; HEL-S-7; LC5; lipocortin V; PAP-I; placental anticoagulant protein 4; Placental anticoagulant protein I; PP4; R74653; RPRGL3; Thromboplastin inhibitor; VAC-alpha; Vascular anticoagulant-alpha
Common Name Annexin V
Gene Symbol Anxa5
Biological Activity Annexin V exhibits anti-phospholipase activity and binds to phosphatidylserine (PS).
Product Type Protein
Specific Reactivity All
Conjugate FITC
Cross Reactivity All
Species E. coli
Recombinant Recombinant
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >89% by SDS-PAGE and RP-HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.