missing translation for 'onlineSavingsMsg'
Learn More

anti-Actin, Clone: C4, MP Biomedicals

Brand:  MP Biomedicals 08691001

Product Code. 11489335

  • ONLINE EXCLUSIVE PRICE
    580.00€
    405.00€ / 0.10mL
    Save 175.00€
    30% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Host Species: Mouse
Clone: C4
Isotype: IgG
Species Reactivity: Human

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Actin
C4
Mouse
Primary
Liquid
Monoclonal
Unconjugated
0.1 mL
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.