Learn More
CD105 (Endoglin) Mouse anti-Human, PE, Clone: 266, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560839
Additional Details : Weight : 0.10000kg
Description
The 266 monoclonal antibody specifically binds to CD105. CD105 presents an integral membrane homodimer protein with subunits of 95 kDa found on vascular endothelial cells and syncytiotrophoblasts of placenta. CD105 is weakly expressed on stromal fibroblasts. It is also expressed on U937 cells, activated macrophages, and mesenchymal stem cells. CD105 is a component of the TGF-β receptor system in human umbilical vein endothelial cells and binds TGF-β1 and TGF-β3 with high affinity but does not bind toTGF-β2. Expression of CD105 is increased on activated endothelium in tissues undergoing angiogenesis, such as in tumors, or in cases of wound healing or dermal inflammation.
Specifications
CD105 | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Umbilical Vein Endothelial Cells | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
266 | |
Endoglin; ENG; END; HHT1; ORW; ORW1 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |