missing translation for 'onlineSavingsMsg'
Learn More

CD121b Rat anti-Mouse, PE, Clone: 4E2, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 554450

Product Code. 15897698

  • ONLINE EXCLUSIVE PRICE
    505.00€
    502.90€ / Each
    Save 2.10€
    0.42% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 4E2 monoclonal antibody specifically binds to CD121b which is also known as the IL-1 receptor type II (IL-1RII, IL-1R2) or IL-1 receptor beta (IL-1R-beta, IL-1Rβ). CD121b is encoded by the Il1r2 gene and presents as a 68 kDa type I transmembrane glycoprotein. It is expressed on a variety of cell types including B cells, neutrophils, monocytes, and bone marrow progenitor cells. A soluble form of CD121b is produced by proteolysis especially after cellular activation. Unlike the IL-1 type I receptor (CD121a), CD121b has a short cytoplasmic domain and does not mediate IL-1 signaling. CD121b may function in a membrane or soluble form as a decoy receptor that can bind IL-1 and reduce its effective concentration and biological activity. The 4E2 antibody blocks IL-1 binding to the IL-1 receptor type II.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD121b
Monoclonal
0.2mg/mL
Il1r2; IL-1RII; IL-1R2; Il1rb; IL-1Rβ; IL-1R-beta; IL-1 receptor beta chain
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
4.00E+02
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse IL-1RII Protein
0.2 mg
Immunology
Mouse
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.