missing translation for 'onlineSavingsMsg'
Learn More

CD15 Mouse anti-Human, FITC, Clone: HI98 (also known as HIM1), BD

Mouse Monoclonal Antibody

84.55€ - 180.52€

Specifications

Antigen CD15
Clone HI98 (also known as HIM1)
Applications Flow Cytometry
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15880669
View Documents
BD Biosciences
560997
25 Tests
84.55€
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15848138
View Documents
BD Biosciences
555401
100 Tests
ONLINE EXCLUSIVE PRICE
181.00€
180.52€ / Each
Save 0.48€
0.27% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reacts with 3-fucosyl-N-acetyllactosamine (3-FAL), a 220kDa carbohydrate structure, also called X-hapten, SSEA-1, CD15 or Lewis X. 3-FAL is expressed on >95% of granulocytes, including neutrophils and eosinophils, and to a varying degree on monocytes, but not on lymphocytes or basophils. CD15 plays a role in mediating phagocytosis, bactericidal activity and chemotaxis. This antibody is also suitable for staining formalin-fixed, paraffin-embedded tissue sections without pretreatment. Since the Abs are recognizing a carbohydrate epitope (3-fucosyl-N-acetyllactosamine) they also should work across species and not only for human.

Flow Cytometry

Specifications

Specifications

CD15
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgM κ
Affinity Purified
HI98 (also known as HIM1)
Monoclonal
Mouse
Human
Human Mononuclear Cells
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.