Learn More
CD20 Mouse anti-Human, PE-CF594, Clone: 2H7, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562322
Description
The 2H7 monoclonal antibody specifically binds to CD20 that is encoded by the MS4A1 (Membrane-spanning 4-domains, subfamily A, member 1) gene. CD20 is a 33-37 kDa unglycosylated four-transmembrane phosphoprotein. CD20 is expressed on pre-B-cells, resting and activated B cells and follicular dendritic cells but not on plasma cells. Low level CD20 expression is observed on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.
Specifications
| CD20 | |
| Monoclonal | |
| PE-CF594 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human 6.16c1.3 B cell line | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 2H7 | |
| MS4A1; B1; Bp35; LEU-16; S7 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.