missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD23; Clone: B3B4; BD
Brand: BD Biosciences 562826
Additional Details : Weight : 0.01000kg
Description
Host Species: Rat
Clone B3B4
Isotype IgG2a, κ
Species Reactivity Ms
Immunogen FcΕR isolated from the mouse B hybridoma line O1.2B2
Flow Cytometry
Specifications
CD23 | |
B3B4 | |
Alexa Fluor 647 | |
FcΕR isolated from the mouse B hybridoma line O1.2B2 | |
100 μg | |
Primary | |
IgG2a |
Monoclonal | |
0.2mg/mL | |
Rat | |
Affinity Purified | |
RUO | |
Mouse |