missing translation for 'onlineSavingsMsg'
Learn More

CD25 Mouse anti-Human, Brilliant Violet 650, Clone: M-A251, BD

Mouse Monoclonal Antibody

231.67€ - 484.68€

Specifications

Antigen CD25
Clone M-A251
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Violet 650
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15853029
View Documents
BD Biosciences
563718
25 Tests
ONLINE EXCLUSIVE PRICE
232.00€
231.67€ / Each
Save 0.33€
0.14% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15863029
View Documents
BD Biosciences
563719
100 Tests
ONLINE EXCLUSIVE PRICE
485.00€
484.68€ / Each
Save 0.32€
0.07% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The M-A251 monoclonal antibody specifically binds to the 55 kDa type I transmembrane glycoprotein known as the low-affinity interleukin-2 receptor alpha chain subunit (IL-2R α ). CD25 is expressed on regulatory T cells and on activated lymphocytes (T and B) and monocytes. It associates with the IL-2R β /CD122 and the IL-2R γ/CD132 receptor chains to form the high-affinity IL-2R complex. CD25 expression on T and B lymphocytes is upregulated by antigenic or mitogenic stimulation. Soluble CD25/IL-2R α is produced as a consequence of lymphocyte stimulation and is found in biological fluids following inflammatory responses.

Specifications

Specifications

CD25
Flow Cytometry
Brilliant Violet 650
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
M-A251
Monoclonal
Mouse
Human
IgG1 κ
Affinity Purified
IL-2R; IL2RA; IL-2Rα; TCGFR; TAC antigen; p55
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.