Learn More
CD300f Monoclonal Antibody (234903), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA523943
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, no cross-reactivity with recombinant human LMIR1, 2, 4, 5, 6, or recombinant mouse LMIR3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
CD300f Monoclonal antibody specifically detects CD300f in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
CD300f | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8TDQ1 | |
CD300LF | |
Y3 rat myeloid cell line transfected with human CD300f/LMIR3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry | |
234903 | |
Unconjugated | |
CD300LF | |
CD300 antigen like family member F; CD300 antigen-like family member F; CD300 molecule like family member f; CD300 molecule-like family member f; CD300f; Cd300lf; CLIM1; CLM1; CLM-1; CMRF-35-like molecule 1; CMRF35-like molecule 1; DC-derived Ig-like receptor 2; Digr2; F730004D16Rik; IgSF13; Immune receptor expressed on myeloid cells 1; immunoglobin superfamily member 13; immunoglobulin superfamily member 13; inhibitory receptor IREM1; IREM1; IREM-1; Leukocyte mono-Ig-like receptor 3; Lmir3; MAIR-5; MAIR-V; myeloid-associated immunoglobulin-like receptor 5; NK inhibitory receptor; NKIR; Pigr3; UNQ3105/PRO10111 | |
Rat | |
Protein A/G | |
RUO | |
146722 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |