missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD33; Clone: WM53; BD
Brand: BD Biosciences 563257
Additional Details : Weight : 0.00820kg
Description
Host Species: Mouse
Clone WM53
Isotype IgG1, κ
Species Reactivity Hu
Immunogen Acute Myeloid Leukemia Blasts
Flow Cytometry
Specifications
CD33 | |
WM53 | |
Mouse | |
Affinity Purified | |
RUO | |
Human |
Monoclonal | |
Brilliant Violet 510 | |
Acute Myeloid Leukemia Blasts | |
50 Tests | |
Primary | |
IgG1 |